Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pnrc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026383.3 | NP_080659.1 | 1 - 2 | 175 | 94 | |
GenBank mRNA | AK011192.1 | - | 1 - 2 | 153 | 94 | |
AK012073.1 | - | 1 - 2 | 159 | 94 | ||
AK018384.1 | - | 1 - 2 | 123 | 94 | ||
BC006598.1 | - | 1 - 2 | 148 | 3965195 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |