Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cnpy4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178612.4 | NP_848727.1 | 2 - 3 | 314 | 76 | |
GenBank mRNA | AK011474.1 | - | 2 - 3 | 315 | 76 | |
AK012968.1 | - | 2 - 3 | 296 | 76 | ||
AK033219.1 | - | 2 - 3 | 316 | 76 | ||
AK051562.1 | - | 2 - 3 | 316 | 76 | ||
BC086647.1 | - | 2 - 3 | 279 | 6818372 | 76 | |
BC094672.1 | - | 2 - 3 | 421 | 30633103 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |