Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Foxk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199068.2 | NP_951031.2 | 3 - 4 | 888 | 66 | |
GenBank mRNA | AK144418.1 | - | 3 - 4 | 781 | 66 | |
AK147375.1 | - | 3 - 4 | 886 | 66 | ||
BC060238.1 | - | 3 - 4 | 868 | 66 | ||
L26507.1 | - | 3 - 4 | 864 | 66 | ||
U95016.1 | - | 3 - 4 | 864 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |