Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbx3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011535.3 | NP_035665.2 | 4 - 5 | 1644 | 95 | |
NM_198052.2 | NP_932169.1 | 4 - 5 | 1780 | 95 | ||
GenBank mRNA | AK052453.1 | - | 4 - 5 | 1859 | 95 | |
AK157247.1 | - | 3 - 4 | 1345 | 95 | ||
AK172118.1 | - | 3 - 4 | 1347 | 95 | ||
BC096551.1 | - | 4 - 5 | 1780 | 30547736 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |