Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lin54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001115010.1 | NP_001108482.1 | 2 - 3 | 994 | 89 | |
NM_172714.4 | NP_766302.2 | 2 - 3 | 862 | 89 | ||
GenBank mRNA | AK030894.1 | - | 2 - 3 | 996 | 89 | |
AK134320.1 | - | 2 - 3 | 860 | 89 | ||
AK220225.1 | - | 2 - 3 | 857 | 89 | ||
BC055819.1 | - | 1 - 2 | 279 | 89 | ||
BC106126.1 | - | 1 - 2 | 498 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |