Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttc28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001267622.1 | NP_001254551.1 | 19 - 20 | 5635 | 70 | |
GenBank mRNA | AK031458.1 | - | 2 - 3 | 229 | 70 | |
AK081382.1 | - | 2 - 3 | 107 | 70 | ||
BC046779.1 | - | 13 - 14 | 3299 | 5719497 | 70 | |
BC060735.1 | - | 12 - 13 | 2295 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |