Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clu | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013492.2 | NP_038520.2 | 6 - 7 | 1165 | 60 | |
GenBank mRNA | AF248057.1 | - | 6 - 7 | 1048 | 60 | |
AF248058.1 | - | 3 - 4 | 719 | 60 | ||
BC075668.1 | - | 6 - 7 | 994 | 30550773 | 60 | |
D14077.1 | - | 6 - 7 | 1023 | 60 | ||
L05670.1 | - | 6 - 7 | 1023 | 60 | ||
L08235.1 | - | 5 - 6 | 995 | 60 | ||
S70244.1 | - | 5 - 6 | 959 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |