Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abcc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013806.2 | NP_038834.2 | 15 - 16 | 2060 | 69 | |
GenBank mRNA | AF227274.1 | - | 15 - 16 | 1964 | 69 | |
AF282772.1 | - | 15 - 16 | 1964 | 69 | ||
AF282773.1 | - | 15 - 16 | 1964 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Detect Genomic DNA Mus musculus Probe spans exons |