Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lig1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083188.1 | NP_001076657.1 | 14 - 15 | 1591 | 96 | |
NM_001199310.1 | NP_001186239.1 | 14 - 15 | 1599 | 96 | ||
NM_010715.2 | NP_034845.2 | 14 - 15 | 1516 | 96 | ||
GenBank mRNA | AK153993.1 | - | 14 - 15 | 1478 | 96 | |
AK161793.1 | - | 7 - 8 | 674 | 96 | ||
AK168643.1 | - | 14 - 15 | 1593 | 96 | ||
BC028287.1 | - | 14 - 15 | 1441 | 4911261 | 96 | |
BC138541.1 | - | 14 - 15 | 1599 | 8861561 | 96 | |
BC138542.1 | - | 14 - 15 | 1599 | 8861562 | 96 | |
U04674.1 | - | 13 - 14 | 1384 | 96 | ||
U19604.1 | - | 14 - 15 | 1629 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |