Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trappc9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029640.2 | NP_083916.1 | 20 - 21 | 2542 | 95 | |
NM_180662.2 | NP_850993.2 | 20 - 21 | 3287 | 95 | ||
GenBank mRNA | AK007766.1 | - | 3 - 4 | 322 | 95 | |
AK156739.1 | - | 21 - 22 | 3112 | 95 | ||
AK173287.1 | - | 10 - 11 | 1286 | 95 | ||
AY630620.1 | - | 20 - 21 | 2540 | 95 | ||
BC019994.1 | - | 6 - 7 | 772 | 95 | ||
BC034590.1 | - | 20 - 21 | 2540 | 5364921 | 95 | |
BC038938.1 | - | 14 - 15 | 1857 | 95 | ||
BC070463.1 | - | 20 - 21 | 3287 | 30613261 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |