Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snrnp70 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK005045.2 | - | 2 - 3 | 339 | 65 | |
AK011564.1 | - | 2 - 3 | 295 | 65 | ||
AK014311.2 | - | 2 - 3 | 336 | 65 | ||
AK077425.1 | - | 2 - 3 | 341 | 65 | ||
AK133115.1 | - | 2 - 3 | 340 | 65 | ||
AK146729.1 | - | 2 - 3 | 340 | 65 | ||
AK148013.1 | - | 2 - 3 | 366 | 65 | ||
BC049128.1 | - | 2 - 3 | 305 | 65 | ||
BC051414.1 | - | 1 - 2 | 140 | 65 | ||
BC131994.1 | - | 2 - 3 | 232 | 40130271 | 65 | |
BC132020.1 | - | 2 - 3 | 232 | 40130297 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |