Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spty2d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175318.4 | NP_780527.2 | 1 - 2 | 338 | 60 | |
GenBank mRNA | AK030869.1 | - | 1 - 2 | 173 | 60 | |
AK147856.1 | - | 1 - 2 | 147 | 60 | ||
AK162467.1 | - | 1 - 2 | 153 | 60 | ||
BC079859.1 | - | 1 - 2 | 133 | 6834544 | 60 | |
BY787796.1 | - | 1 - 2 | 340 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |