Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Med25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029365.2 | NP_083641.1 | 14 - 15 | 1673 | 89 | |
GenBank mRNA | AK011662.1 | - | 5 - 6 | 572 | 89 | |
AK089940.1 | - | 14 - 15 | 1677 | 89 | ||
AK090026.1 | - | 14 - 15 | 1700 | 89 | ||
AK165760.1 | - | 14 - 15 | 90 | |||
BC021333.1 | - | 14 - 15 | 1708 | 5027401 | 89 | |
BC031138.1 | - | 14 - 15 | 1716 | 4972378 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |