Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trpm4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175130.4 | NP_780339.2 | 3 - 4 | 335 | 68 | |
GenBank mRNA | AJ575814.1 | - | 3 - 4 | 314 | 68 | |
BC096475.1 | - | 3 - 4 | 311 | 4225167 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Ion Channel Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |