Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mamdc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174857.3 | NP_777282.1 | 10 - 11 | 1763 | 120 | |
GenBank mRNA | AK004794.1 | - | 9 - 10 | 1626 | 120 | |
AK035566.1 | - | 10 - 11 | 1764 | 120 | ||
AK154219.1 | - | 10 - 11 | 1840 | 120 | ||
BC042773.1 | - | 10 - 11 | 1792 | 4019075 | 120 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |