Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnah10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019536.1 | NP_062409.1 | 50 - 51 | 8717 | 94 | |
GenBank mRNA | AK147443.1 | - | 3 - 4 | 681 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Mus musculus Probe spans exons |