Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Shank1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034115.1 | NP_001029287.1 | 5 - 6 | 716 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Mus musculus Probe spans exons |