Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gtf2ird2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053266.1 | NP_444496.1 | 7 - 8 | 846 | 62 | |
GenBank mRNA | AK082488.1 | - | 1015 | 62 | ||
AY014963.1 | - | 7 - 8 | 846 | 62 | ||
AY116023.1 | - | 7 - 8 | 857 | 62 | ||
BC057179.1 | - | 7 - 8 | 668 | 62 | ||
BC132157.1 | - | 7 - 8 | 754 | 40130434 | 62 | |
BC132159.1 | - | 7 - 8 | 754 | 40130436 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |