Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdcd1lg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021396.2 | NP_067371.1 | 3 - 4 | 601 | 68 | |
GenBank mRNA | AF142780.2 | - | 3 - 4 | 574 | 68 | |
AK089369.1 | - | 3 - 4 | 604 | 68 | ||
AK141111.1 | - | 2 - 3 | 691 | 68 | ||
AK155002.1 | - | 3 - 4 | 541 | 68 | ||
BC104138.1 | - | 3 - 4 | 506 | 40045578 | 68 | |
BC104139.1 | - | 3 - 4 | 506 | 40045580 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |