Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wapal | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004436.4 | NP_001004436.2 | 5 - 6 | 2217 | 100 | |
NM_001301330.1 | NP_001288259.1 | 5 - 6 | 2199 | 100 | ||
GenBank mRNA | AB167349.1 | - | 5 - 6 | 2023 | 100 | |
AK028167.1 | - | 3 - 4 | 321 | 100 | ||
AK129105.1 | - | 5 - 6 | 2217 | 100 | ||
BC139434.1 | - | 5 - 6 | 1972 | 8862456 | 100 | |
BC145625.1 | - | 5 - 6 | 1954 | 9054193 | 100 | |
CF161743.1 | - | 3 - 4 | 324 | 100 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |