Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sncg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011430.3 | NP_035560.1 | 1 - 2 | 207 | 72 | |
GenBank mRNA | AA690669.1 | - | 1 - 2 | 321 | 72 | |
AF017255.1 | - | 1 - 2 | 186 | 72 | ||
AJ786826.1 | - | 1 - 2 | 120 | 72 | ||
AK141509.1 | - | 1 - 2 | 191 | 72 | ||
BC028508.1 | - | 1 - 2 | 198 | 1448798 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |