Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gyk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001294138.1 | NP_001281067.1 | 1 - 2 | 385 | 149 | |
NM_001294140.1 | NP_001281069.1 | 1 - 2 | 385 | 149 | ||
NM_008194.3 | NP_032220.1 | 1 - 2 | 385 | 149 | ||
NM_212444.2 | NP_997609.1 | 1 - 2 | 385 | 149 | ||
GenBank mRNA | AK008186.1 | - | 1 - 2 | 189 | 149 | |
AK044308.1 | - | 1 - 2 | 223 | 149 | ||
AK088373.1 | - | 1 - 2 | 204 | 149 | ||
AK169424.1 | - | 1 - 2 | 191 | 149 | ||
AK169525.1 | - | 1 - 2 | 386 | 149 | ||
BC003767.1 | - | 1 - 2 | 188 | 3593896 | 149 | |
U48403.1 | - | 1 - 2 | 162 | 149 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |