Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Parg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011960.2 | NP_036090.2 | 5 - 6 | 1834 | 139 | |
GenBank mRNA | AF079557.1 | - | 5 - 6 | 1583 | 139 | |
AK036656.1 | - | 5 - 6 | 1857 | 139 | ||
AK088708.1 | - | 2 - 3 | 189 | 139 | ||
AK160742.1 | - | 4 - 5 | 1308 | 139 | ||
AK164013.1 | - | 5 - 6 | 1821 | 139 | ||
BC050892.1 | - | 5 - 6 | 1833 | 139 | ||
BC053037.1 | - | 5 - 6 | 1817 | 139 | ||
BC059827.1 | - | 5 - 6 | 1843 | 139 | ||
EF382673.1 | - | 3 - 4 | 543 | 139 | ||
JQ890227.1 | - | 2 - 3 | 340 | 139 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |