Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnah12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK076605.1 | - | 6 - 7 | 598 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Mus musculus Probe spans exons |