Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uty | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009484.2 | NP_033510.2 | 4 - 5 | 556 | 121 | |
GenBank mRNA | AF057367.1 | - | 4 - 5 | 536 | 121 | |
AK142344.1 | - | 6 - 7 | 644 | 121 | ||
AK142512.1 | - | 4 - 5 | 472 | 121 | ||
AK155922.1 | - | 5 - 6 | 436 | 121 | ||
BC053061.1 | - | 4 - 5 | 539 | 121 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |