Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nfxl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133921.2 | NP_598682.2 | 20 - 21 | 2500 | 137 | |
GenBank mRNA | AK132320.1 | - | 19 - 20 | 2410 | 137 | |
BC022652.1 | - | 4 - 5 | 500 | 137 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |