Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itih1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008406.3 | NP_032432.2 | 4 - 5 | 429 | 88 | |
GenBank mRNA | AK149500.1 | - | 4 - 5 | 430 | 88 | |
BC013465.1 | - | 4 - 5 | 413 | 88 | ||
BC028814.1 | - | 4 - 5 | 429 | 5099284 | 88 | |
X70391.1 | - | 4 - 5 | 409 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |