Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009441.2 | NP_033467.2 | 1 - 2 | 124 | 70 | |
GenBank mRNA | AK088273.1 | - | 1 - 2 | 125 | 70 | |
AK089974.1 | - | 1 - 2 | 67 | 70 | ||
AK166298.1 | - | 1 - 2 | 66 | 70 | ||
AK170087.1 | - | 1 - 2 | 115 | 70 | ||
BC019173.1 | - | 1 - 2 | 93 | 70 | ||
BC037654.1 | - | 1 - 2 | 69 | 70 | ||
BC049277.1 | - | 1 - 2 | 106 | 70 | ||
BC057207.1 | - | 1 - 2 | 82 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |