Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lmna | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002011.3 | NP_001002011.2 | 7 - 8 | 1634 | 57 | |
NM_001111102.2 | NP_001104572.1 | 7 - 8 | 1634 | 57 | ||
NM_019390.3 | NP_062263.1 | 7 - 8 | 1105 | 57 | ||
GenBank mRNA | AK004619.2 | - | 7 - 8 | 1589 | 57 | |
AK147150.1 | - | 7 - 8 | 1590 | 57 | ||
AK149998.1 | - | 7 - 8 | 1590 | 57 | ||
AK150391.1 | - | 7 - 8 | 1590 | 57 | ||
AK150501.1 | - | 7 - 8 | 1590 | 57 | ||
AK150624.1 | - | 7 - 8 | 1590 | 57 | ||
AK152539.1 | - | 7 - 8 | 1590 | 57 | ||
AK152646.1 | - | 7 - 8 | 1590 | 57 | ||
AK152846.1 | - | 7 - 8 | 1590 | 57 | ||
AK153295.1 | - | 7 - 8 | 1531 | 57 | ||
AK167858.1 | - | 7 - 8 | 1587 | 57 | ||
AK208885.1 | - | 1 - 2 | 29 | 57 | ||
AK216120.1 | - | 1 - 2 | 29 | 57 | ||
BC015302.1 | - | 7 - 8 | 1510 | 4240057 | 57 | |
BC094020.1 | - | 7 - 8 | 1496 | 4187374 | 57 | |
D13181.1 | - | 4 - 5 | 683 | 57 | ||
D14850.1 | - | 7 - 8 | 1105 | 57 | ||
DQ832702.1 | - | 7 - 8 | 1385 | 57 | ||
DQ832703.1 | - | 7 - 8 | 1385 | 57 | ||
X14170.1 | - | 7 - 8 | 1579 | 57 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |