Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif2b4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127355.1 | NP_001120827.1 | 10 - 11 | 1280 | 106 | |
NM_001127356.1 | NP_001120828.1 | 9 - 10 | 1502 | 106 | ||
NM_010122.2 | NP_034252.2 | 10 - 11 | 1135 | 106 | ||
GenBank mRNA | AK154364.1 | - | 10 - 11 | 1043 | 106 | |
AK158282.1 | - | 10 - 11 | 1044 | 106 | ||
BC021884.1 | - | 10 - 11 | 1022 | 4168186 | 106 | |
M98035.1 | - | 9 - 10 | 1122 | 106 | ||
M98036.1 | - | 10 - 11 | 1088 | 106 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |