Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ociad1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159887.1 | NP_001153359.1 | 3 - 4 | 333 | 151 | |
NM_001159888.1 | NP_001153360.1 | 3 - 4 | 333 | 151 | ||
NM_001159889.1 | NP_001153361.1 | 3 - 4 | 333 | 151 | ||
NM_023429.4 | NP_075918.1 | 3 - 4 | 333 | 151 | ||
GenBank mRNA | AF295357.1 | - | 3 - 4 | 284 | 151 | |
AK006013.1 | - | 3 - 4 | 325 | 151 | ||
AK007636.1 | - | 3 - 4 | 317 | 151 | ||
AK043603.1 | - | 3 - 4 | 302 | 151 | ||
AK077904.1 | - | 3 - 4 | 303 | 151 | ||
AK078102.1 | - | 3 - 4 | 334 | 151 | ||
AK078909.1 | - | 3 - 4 | 297 | 151 | ||
AK080795.1 | - | 3 - 4 | 325 | 151 | ||
AK082769.1 | - | 3 - 4 | 327 | 151 | ||
AK082781.1 | - | 3 - 4 | 276 | 151 | ||
AK145907.1 | - | 3 - 4 | 335 | 151 | ||
AK159811.1 | - | 3 - 4 | 325 | 151 | ||
AK167320.1 | - | 3 - 4 | 297 | 151 | ||
AK169715.1 | - | 3 - 4 | 315 | 151 | ||
AK177352.1 | - | 2 - 3 | 167 | 151 | ||
AK177999.1 | - | 1 - 2 | 77 | 151 | ||
AK213268.1 | - | 1 - 2 | 77 | 151 | ||
AY208935.1 | - | 3 - 4 | 232 | 151 | ||
BC006937.1 | - | 3 - 4 | 240 | 2648369 | 151 | |
CX238085.1 | - | 3 - 4 | 281 | 151 | ||
CX238454.1 | - | 3 - 4 | 267 | 151 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |