Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009724.2 | NP_033854.1 | 2 - 3 | 262 | 63 | |
GenBank mRNA | AK008180.1 | - | 2 - 3 | 261 | 63 | |
AK008686.1 | - | 2 - 3 | 262 | 63 | ||
AK009499.1 | - | 2 - 3 | 262 | 63 | ||
AK009670.1 | - | 2 - 3 | 267 | 63 | ||
AK009832.1 | - | 2 - 3 | 264 | 63 | ||
AK009994.1 | - | 2 - 3 | 261 | 63 | ||
AK010003.1 | - | 2 - 3 | 261 | 63 | ||
AK010030.1 | - | 2 - 3 | 262 | 63 | ||
BC120705.1 | - | 2 - 3 | 261 | 40129628 | 63 | |
BC125292.1 | - | 2 - 3 | 261 | 40129807 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |