Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myo16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081397.1 | NP_001074866.1 | 31 - 32 | 4324 | 133 | |
GenBank mRNA | AB429291.1 | - | 28 - 29 | 3830 | 133 | |
AK034257.1 | - | 8 - 9 | 1196 | 133 | ||
AK220356.1 | - | 12 - 13 | 1756 | 133 | ||
BC082311.1 | - | 31 - 32 | 4190 | 133 | ||
BC151049.1 | - | 31 - 32 | 4278 | 9087962 | 133 | |
BC151051.1 | - | 31 - 32 | 4279 | 9087964 | 133 | |
BC157966.1 | - | 31 - 32 | 4279 | 9007290 | 133 | |
BC172122.1 | - | 31 - 32 | 4279 | 9054408 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |