Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pax2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011037.4 | NP_035167.4 | 4 - 5 | 491 | 129 | |
GenBank mRNA | BC148232.1 | - | 4 - 5 | 495 | 40142573 | 129 |
BC150484.1 | - | 4 - 5 | 495 | 40142579 | 129 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |