Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fcrl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113238.1 | NP_001106709.1 | 3 - 4 | 83 | ||
NM_183222.3 | NP_899045.3 | 4 - 5 | 83 | |||
GenBank mRNA | AK089756.1 | - | 3 - 4 | 83 | ||
AY158090.1 | - | 4 - 5 | 707 | 83 | ||
AY506558.1 | - | 4 - 5 | 716 | 83 | ||
BC137997.1 | - | 4 - 5 | 644 | 8861016 | 83 | |
BC137998.1 | - | 4 - 5 | 644 | 8861017 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |