Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tdo2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019911.2 | NP_064295.2 | 7 - 8 | 763 | 135 | |
GenBank mRNA | AB476989.1 | - | 7 - 8 | 1237 | 135 | |
AB476990.1 | - | 7 - 8 | 1123 | 135 | ||
AK132271.1 | - | 7 - 8 | 786 | 135 | ||
BC018390.1 | - | 7 - 8 | 763 | 4159877 | 135 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |