Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wfs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011716.2 | NP_035846.1 | 3 - 4 | 440 | 55 | |
GenBank mRNA | AF084482.1 | - | 3 - 4 | 373 | 55 | |
AJ011971.1 | - | 3 - 4 | 367 | 55 | ||
AK144561.1 | - | 3 - 4 | 392 | 55 | ||
AK145023.1 | - | 3 - 4 | 412 | 55 | ||
AK170180.1 | - | 3 - 4 | 395 | 55 | ||
BC046988.1 | - | 3 - 4 | 376 | 6492038 | 55 | |
CX562372.1 | - | 1 - 2 | 70 | 55 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |