Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zbbx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172515.2 | NP_766103.2 | 8 - 9 | 960 | 82 | |
GenBank mRNA | AK029899.1 | - | 8 - 9 | 953 | 82 | |
AK133102.1 | - | 8 - 9 | 1011 | 82 | ||
BC050789.1 | - | 8 - 9 | 960 | 6706914 | 82 | |
BC100435.1 | - | 8 - 9 | 960 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |