Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itih3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008407.2 | NP_032433.2 | 20 - 21 | 2467 | 77 | |
GenBank mRNA | AK146700.1 | - | 19 - 20 | 2956 | 77 | |
BC015276.1 | - | 20 - 21 | 2425 | 4212290 | 77 | |
X70393.1 | - | 19 - 20 | 2424 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |