Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hapln2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022031.2 | NP_071314.1 | 1 - 2 | 127 | 110 | |
GenBank mRNA | AB049055.1 | - | 1 - 2 | 56 | 110 | |
AK081264.1 | - | 1 - 2 | 108 | 110 | ||
BY122070.1 | - | 1 - 2 | 128 | 110 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Mus musculus Probe spans exons |