Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nes | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016701.3 | NP_057910.3 | 3 - 4 | 1097 | 96 | |
GenBank mRNA | AF076623.2 | - | 3 - 4 | 1061 | 96 | |
AK009706.1 | - | 3 - 4 | 1002 | 96 | ||
AK031207.1 | - | 3 - 4 | 1123 | 96 | ||
BC060693.1 | - | 3 - 4 | 1080 | 6410072 | 96 | |
BC062893.1 | - | 3 - 4 | 1097 | 6405026 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Inventoried Probe spans exons |