Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Maged1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019791.2 | NP_062765.1 | 7 - 8 | 1858 | 97 | |
GenBank mRNA | AB029448.1 | - | 7 - 8 | 1858 | 97 | |
AF319975.1 | - | 6 - 7 | 1628 | 97 | ||
AK017275.1 | - | 7 - 8 | 1776 | 97 | ||
AK049660.1 | - | 7 - 8 | 1809 | 97 | ||
AK087937.1 | - | 7 - 8 | 1808 | 97 | ||
AK132248.1 | - | 7 - 8 | 1797 | 97 | ||
AK135452.1 | - | 7 - 8 | 1808 | 97 | ||
AK145872.1 | - | 7 - 8 | 1778 | 97 | ||
AK146867.1 | - | 7 - 8 | 1808 | 97 | ||
AK146908.1 | - | 7 - 8 | 1808 | 97 | ||
AK150876.1 | - | 4 - 5 | 797 | 97 | ||
AK195822.1 | - | 4 - 5 | 270 | 97 | ||
AK220150.1 | - | 8 - 9 | 1715 | 97 | ||
BC031461.1 | - | 7 - 8 | 1798 | 4018875 | 97 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Apoptosis Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |