Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eml4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114361.1 | NP_001107833.1 | 17 - 18 | 2215 | 86 | |
NM_001114362.1 | NP_001107834.1 | 15 - 16 | 2008 | 86 | ||
NM_001286567.1 | NP_001273496.1 | 17 - 18 | 2118 | 86 | ||
NM_199466.3 | NP_955760.3 | 14 - 15 | 1879 | 86 | ||
GenBank mRNA | AK139067.1 | - | 1 - 2 | 72 | 86 | |
AK144604.1 | - | 17 - 18 | 2216 | 86 | ||
BC032258.1 | - | 3 - 4 | 363 | 86 | ||
BC065178.1 | - | 17 - 18 | 2056 | 6827397 | 86 | |
BC066099.1 | - | 14 - 15 | 1810 | 30533573 | 86 | |
BC067011.1 | - | 14 - 15 | 1828 | 6405184 | 86 | |
BC070427.1 | - | 15 - 16 | 1948 | 6413041 | 86 | |
BC079619.1 | - | 14 - 15 | 1810 | 30533573 | 86 | |
DQ230326.1 | - | 16 - 17 | 1898 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |