Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mgat4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290801.1 | NP_001277730.1 | 8 - 9 | 1129 | 142 | |
NM_173870.3 | NP_776295.1 | 9 - 10 | 1208 | 142 | ||
GenBank mRNA | AB053217.1 | - | 8 - 9 | 884 | 142 | |
AK052278.1 | - | 6 - 7 | 705 | 142 | ||
AK155724.1 | - | 9 - 10 | 1209 | 142 | ||
AK163327.1 | - | 8 - 9 | 961 | 142 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |