Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rev1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019570.3 | NP_062516.2 | 9 - 10 | 1749 | 99 | |
GenBank mRNA | AB057418.1 | - | 8 - 9 | 1538 | 99 | |
AF179302.1 | - | 8 - 9 | 1538 | 99 | ||
AK048838.1 | - | 9 - 10 | 1754 | 99 | ||
BC058093.1 | - | 9 - 10 | 1749 | 6827108 | 99 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |