Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mst1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008243.3 | NP_032269.3 | 14 - 15 | 1643 | 82 | |
GenBank mRNA | AK040934.1 | - | 14 - 15 | 1627 | 82 | |
AK134222.1 | - | 14 - 15 | 1626 | 82 | ||
AK144824.1 | - | 14 - 15 | 1606 | 82 | ||
BC010551.1 | - | 14 - 15 | 1628 | 4195560 | 82 | |
BC051393.1 | - | 14 - 15 | 1612 | 5055055 | 82 | |
M74181.1 | - | 14 - 15 | 1600 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |