Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbm6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011251.3 | NP_035381.2 | 16 - 17 | 2911 | 82 | |
NM_029169.3 | NP_083445.1 | 16 - 17 | 3182 | 82 | ||
GenBank mRNA | AJ006486.2 | - | 16 - 17 | 2790 | 82 | |
AK013915.1 | - | 7 - 8 | 604 | 82 | ||
AK077899.1 | - | 16 - 17 | 3089 | 82 | ||
AK145612.1 | - | 16 - 17 | 3071 | 82 | ||
AK164432.1 | - | 11 - 12 | 1268 | 82 | ||
AK220446.1 | - | 16 - 17 | 3060 | 82 | ||
BC026130.1 | - | 16 - 17 | 3085 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |