Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tcirg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001136091.2 | NP_001129563.1 | 10 - 11 | 1288 | 79 | |
NM_001167784.1 | NP_001161256.1 | 10 - 11 | 1633 | 79 | ||
NM_016921.3 | NP_058617.3 | 10 - 11 | 1329 | 79 | ||
GenBank mRNA | AB022322.1 | - | 10 - 11 | 1195 | 79 | |
AF218253.1 | - | 10 - 11 | 1267 | 79 | ||
AF218254.1 | - | 4 - 5 | 871 | 79 | ||
AK004094.1 | - | 1 - 2 | 139 | 79 | ||
AK155140.1 | - | 10 - 11 | 1634 | 79 | ||
BC006761.1 | - | 10 - 11 | 1329 | 3257437 | 79 | |
BC085234.1 | - | 10 - 11 | 1189 | 5322429 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |