Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ehbp1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114595.1 | NP_001108067.1 | 8 - 9 | 1043 | 103 | |
NM_001114596.1 | NP_001108068.1 | 9 - 10 | 1079 | 103 | ||
GenBank mRNA | AF305089.1 | - | 8 - 9 | 996 | 103 | |
AF305090.1 | - | 9 - 10 | 1032 | 103 | ||
AK194109.1 | - | 3 - 4 | 204 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |