Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cox7a2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159529.1 | NP_001153001.1 | 2 - 3 | 262 | 74 | |
GenBank mRNA | AK009614.1 | - | 2 - 3 | 259 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Transporter Detect Genomic DNA Mus musculus 1700 Probe spans exons |